Anti-DRD1 Mouse Monoclonal Antibody [clone: SG2-D1a]

Supplier: Abnova
MAB2314
10711-270EA 530.32 USD
10711-270
Anti-DRD1 Mouse Monoclonal Antibody [clone: SG2-D1a]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Drd1a.

Type: Primary
Antigen: DRD1
Clonality: Monoclonal
Clone: SG2-D1a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR