Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]

Supplier: Abnova
MAB2342
10720-860EA 530.32 USD
10720-860
Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HIF1A.

Recommended Dilutions: Immunocytochemistry: 1:100-1:5000; Immunofluorescence: 1:100-1:5000; Immunohistochemistry (Frozen sections): 1:100-1:5000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:5000

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: ESEE122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR