Anti-ESPL1 Mouse Monoclonal Antibody [clone: XJ11-4D7]

Supplier: Abnova
MAB2347
10712-456EA 530.32 USD
10712-456
Anti-ESPL1 Mouse Monoclonal Antibody [clone: XJ11-4D7]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ESPL1.

Recommended Dilutions: Immunofluorescence: 1:1000

Type: Primary
Antigen: ESPL1
Clonality: Monoclonal
Clone: XJ11-4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR