Anti-CITED2 Mouse Monoclonal Antibody [clone: JA22]

Supplier: Abnova
MAB2353
10708-010EA 530.32 USD
10708-010
Anti-CITED2 Mouse Monoclonal Antibody [clone: JA22]
Antibodies
Mouse monoclonal antibody raised against CITED2.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CITED2
Clonality: Monoclonal
Clone: JA22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR