Anti-CHAF1A Mouse Monoclonal Antibody [clone: SS 1 1-13]

Supplier: Abnova
MAB2356
10673-748EA 530.32 USD
10673-748
Anti-CHAF1A Mouse Monoclonal Antibody [clone: SS 1 1-13]
Antibodies
Mouse monoclonal antibody raised against recombinant CHAF1A.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CHAF1A
Clonality: Monoclonal
Clone: SS 1 1-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR