Anti-CASP3 Mouse Monoclonal Antibody [clone: CPP32 4-1-18]

Supplier: Abnova
MAB2360
10673-324EA 530.32 USD
10673-324
Anti-CASP3 Mouse Monoclonal Antibody [clone: CPP32 4-1-18]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CASP3.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: CPP32 4-1-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR