Anti-MIS18A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21314
10705-016EA 428.55 USD
10705-016
Anti-MIS18A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C21orf45.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: MIS18A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR