Anti-H-2KD Mouse Monoclonal Antibody [clone: 34-1-2S]

Supplier: Abnova
MAB3259
10735-034EA 345.2 USD
10735-034
Anti-H-2KD Mouse Monoclonal Antibody [clone: 34-1-2S]
Antibodies
Mouse monoclonal antibody raised against H-2Kd/H-2Dd.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: H-2Kd/H-2Dd
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 34-1-2S
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR