Anti-H2-D4 Mouse Monoclonal Antibody [clone: 34-5-8S]

Supplier: Abnova
MAB3260
10735-018EA 329 USD
10735-018
Anti-H2-D4 Mouse Monoclonal Antibody [clone: 34-5-8S]
Antibodies
Mouse monoclonal antibody raised against H2-D4.

Type: Primary
Antigen: H2-D4
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 34-5-8S
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR