Anti-H-2Dk Mouse Monoclonal Antibody [clone: 15-5-5S]

Supplier: Abnova
MAB3262
10735-024EA 329 USD
10735-024
Anti-H-2Dk Mouse Monoclonal Antibody [clone: 15-5-5S]
Antibodies
Mouse monoclonal antibody raised against H-2Dk.

Type: Primary
Antigen: H-2Dk
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 15-5-5S
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR