Anti-LAMP1 Rat Monoclonal Antibody [clone: 1D4B]

Supplier: Abnova
MAB3265
10735-404EA 346 USD
10735-404
Anti-LAMP1 Rat Monoclonal Antibody [clone: 1D4B]
Antibodies
Rat monoclonal antibody raised against native Lamp1.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 1D4B
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR