Anti-Light Chain Mouse Monoclonal Antibody [clone: SB62a]

Supplier: Abnova
MAB3268
10735-610EA 329 USD
10735-610
Anti-Light Chain Mouse Monoclonal Antibody [clone: SB62a]
Antibodies
Mouse monoclonal antibody raised against Rabbit Light Chain.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: Light Chain
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SB62a
Epitope:
Host: IgG2b
Isotype: Mouse
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR