Anti-S tag Monoclonal Antibody [clone: SBSTAGb]

Supplier: Abnova
MAB3272
10735-626EA 638.45 USD
10735-626
Anti-S tag Monoclonal Antibody [clone: SBSTAGb]
Antibodies
Mouse monoclonal antibody raised against recombinant S tag.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: S tag
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SBSTAGb
Epitope:
Host: IgG2a
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR