Anti-MAD2L1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12342
10724-368EA 531 USD
10724-368
Anti-MAD2L1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant MAD2.

Recommended Dilutions: Western Blot: 1:100

Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR