Anti-Bu-1a Mouse Monoclonal Antibody [clone: 21-1A4]

Supplier: Abnova
MAB3275
10734-654EA 345.2 USD
10734-654
Anti-Bu-1a Mouse Monoclonal Antibody [clone: 21-1A4]
Antibodies
Mouse monoclonal antibody raised against Bu-1a.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: Bu-1a
Clonality: Monoclonal
Clone: 21-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR