Anti-Cytomegalovirus Immediate Early Antigen (IEA), pp72 Mouse Monoclonal Antibody [clone: 1-K-10]

Supplier: Abnova
MAB3972
10708-360EA 638.45 USD
10708-360
Anti-Cytomegalovirus Immediate Early Antigen (IEA), pp72 Mouse Monoclonal Antibody [clone: 1-K-10]
Antibodies
Mouse monoclonal antibody raised against Cytomegalovirus, Immediate Early Antigen (IEA), pp72.

Recommended Dilutions: Immunofluorescence: 1:1000; ELISA: 1:10000

Type: Primary
Antigen: Cytomegalovirus Immediate Early Antigen (IEA), pp72
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1-K-10
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR