Anti-Dinitrophenol (DNP) Monoclonal Antibody [clone: B136M]

Supplier: Abnova
MAB3977
10710-714EA 1169.22 USD
10710-714
Anti-Dinitrophenol (DNP) Monoclonal Antibody [clone: B136M]
Antibodies
Mouse monoclonal antibody raised against Dinitrophenol (DNP).

Recommended Dilutions: Recommend dilutions start at 5 ?g/ml.

Type: Primary
Antigen: Dinitrophenol (DNP)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: B136M
Epitope:
Host: IgG1
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR