Anti-FANCG Polyclonal Antibody

Supplier: Abnova
PAB12371
10714-368EA 530.32 USD
10714-368
Anti-FANCG Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FANCG.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FANCG
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR