Anti-AFP Mouse Monoclonal Antibody [clone: 131-1221]

Supplier: Abnova
MAB4007
10626-512EA 638.45 USD
10626-512
Anti-AFP Mouse Monoclonal Antibody [clone: 131-1221]
Antibodies
Mouse monoclonal antibody raised against native AFP.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 131-1221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR