Anti-APOA1 Mouse Monoclonal Antibody [clone: 57-161]

Supplier: Abnova
MAB4011
10626-590EA 428.44 USD
10626-590
Anti-APOA1 Mouse Monoclonal Antibody [clone: 57-161]
Antibodies
Mouse monoclonal antibody raised against APOA1.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 57-161
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR