Anti-APOA1 Mouse Monoclonal Antibody [clone: 144]

Supplier: Abnova
MAB4014
10626-592EA 638.45 USD
10626-592
Anti-APOA1 Mouse Monoclonal Antibody [clone: 144]
Antibodies
Mouse monoclonal antibody raised against native APOA1.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 144
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR