Anti-APOA1 Mouse Monoclonal Antibody [clone: 145]

Supplier: Abnova
MAB4015
10626-538EA 638.45 USD
10626-538
Anti-APOA1 Mouse Monoclonal Antibody [clone: 145]
Antibodies
Mouse monoclonal antibody raised against native APOA1.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: 145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR