Anti-CDCA7L Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21880
10707-194EA 428.55 USD
10707-194
Anti-CDCA7L Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CDCA7L.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: CDCA7L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR