Anti-LRRK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12701
10724-078EA 329 USD
10724-078
Anti-LRRK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LRRK2.

  • Recommended Dilutions: Western Blot: 0.1-1 μg/ml; ELISA: 0.01-0.1 μg/ml; Immunoprecipitation: 2-5 μg/ml

Type: Primary
Antigen: LRRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR