Anti-ZNF114 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22302
10745-492EA 428.55 USD
10745-492
Anti-ZNF114 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ZNF114.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: ZNF114
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR