Anti-GRTP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13368
10720-700EA 428 USD
10720-700
Anti-GRTP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GRTP1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: GRTP1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR