Anti-VPS53 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13370
10744-314EA 428 USD
10744-314
Anti-VPS53 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of VPS53.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: VPS53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR