Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]

Supplier: Abnova
MAB2365
10691-268EA 530.32 USD
10691-268
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 32.1]
Antibodies
Mouse monoclonal antibody raised against full length recombinant BIRC5.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 20 ?g/ml

Type: Primary
Antigen: BIRC5
Clonality: Monoclonal
Clone: 32.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR