Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]

Supplier: Abnova
MAB2377
10692-690EA 530.32 USD
10692-690
Anti-BAP1 Mouse Monoclonal Antibody [clone: 1G8]
Antibodies
Mouse monoclonal antibody raised against full length recombinant BAP1.

Recommended Dilutions: Immunocytochemistry: 1:100; Immunofluorescence: 1:50-1:200; Western Blot: 2 ?g/ml

Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR