Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]

Supplier: Abnova
MAB2382
10688-604EA 530.32 USD
10688-604
Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]
Antibodies
Mouse monoclonal antibody raised against recombinant AIP.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AIP
Clonality: Monoclonal
Clone: 35-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR