Anti-DFFB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13403
10713-254EA 527.69 USD
10713-254
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DFFB.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR