Anti-DFFB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13404
10713-256EA 428 USD
10713-256
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DFFB.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR