Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13411
10744-576EA 428 USD
10744-576
Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Envelope.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: West Nile Virus Envelope
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR