Anti-PTPN5 Mouse Monoclonal Antibody [clone: 2300000]

Supplier: Abnova
MAB2403
10730-686EA 530.32 USD
10730-686
Anti-PTPN5 Mouse Monoclonal Antibody [clone: 2300000]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Ptpn5.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: PTPN5
Clonality: Monoclonal
Clone: 2300000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR