Anti-MURC Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21348
10732-860EA 428.55 USD
10732-860
Anti-MURC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant MURC.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: MURC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR