Anti-TPST2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21351
10742-574EA 428.55 USD
10742-574
Anti-TPST2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TPST2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: TPST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR