Anti-IL5 Rat Monoclonal Antibody [clone: JES1-39D10]

Supplier: Abnova
MAB3295
10735-268EA 346 USD
10735-268
Anti-IL5 Rat Monoclonal Antibody [clone: JES1-39D10]
Antibodies
Rat monoclonal antibody raised against recombinant IL5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: JES1-39D10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR