Anti-IL2 Rat Monoclonal Antibody [clone: JES6-1A12]

Supplier: Abnova
MAB3305
10735-228EA 346 USD
10735-228
Anti-IL2 Rat Monoclonal Antibody [clone: JES6-1A12]
Antibodies
Rat monoclonal antibody raised against recombinant Il2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: JES6-1A12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR