Anti-C9ORF78 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21368
10704-544EA 428.55 USD
10704-544
Anti-C9ORF78 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C9orf78.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: C9ORF78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR