Anti-IL3 Rat Monoclonal Antibody [clone: MP2-8F8]

Supplier: Abnova
MAB3314
10735-260EA 346 USD
10735-260
Anti-IL3 Rat Monoclonal Antibody [clone: MP2-8F8]
Antibodies
Rat monoclonal antibody raised against recombinant Il3.

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: MP2-8F8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR