Anti-CSF2 Rat Monoclonal Antibody [clone: MP1-31G6]

Supplier: Abnova
MAB3321
10734-880EA 346 USD
10734-880
Anti-CSF2 Rat Monoclonal Antibody [clone: MP1-31G6]
Antibodies
Rat monoclonal antibody raised against recombinant Csf2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: MP1-31G6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR