Anti-Benzoylecgonine Monoclonal Antibody [clone: IP3G2]

Supplier: Abnova
MAB4029
10626-728EA 957.05 USD
10626-728
Anti-Benzoylecgonine Monoclonal Antibody [clone: IP3G2]
Antibodies
Mouse monoclonal antibody raised against Benzoylecgonine.

Type: Primary
Antigen: Benzoylecgonine
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: IP3G2
Epitope:
Host: IgG1
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR