Anti-MSR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12378
10726-044EA 530.32 USD
10726-044
Anti-MSR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MSR1.

Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Western Blot: 2 ?g/ml

Type: Primary
Antigen: MSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR