Anti-LRP8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12395
10723-998EA 530.32 USD
10723-998
Anti-LRP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LRP8.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: LRP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR