Anti-PRSS38 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21905
10730-328EA 428.55 USD
10730-328
Anti-PRSS38 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PRSS38.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: PRSS38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR