Anti-LPIN1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12399
10723-888EA 530.32 USD
10723-888
Anti-LPIN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LPIN1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:250

Type: Primary
Antigen: LPIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR