Anti-Ferritin Mouse Monoclonal Antibody [clone: 090-10187]

Supplier: Abnova
MAB4061
10627-316EA 428.44 USD
10627-316
Anti-Ferritin Mouse Monoclonal Antibody [clone: 090-10187]
Antibodies
Mouse monoclonal antibody raised against native Ferritin.

Type: Primary
Antigen: Ferritin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 090-10187
Epitope:
Host: IgG2a kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR