Anti-NOL12 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21911
10727-366EA 428.55 USD
10727-366
Anti-NOL12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant NOL12.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500

Type: Primary
Antigen: NOL12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR