Anti-HBsAg Mouse Monoclonal Antibody [clone: BID40]

Supplier: Abnova
MAB4070
10627-632EA 638.45 USD
10627-632
Anti-HBsAg Mouse Monoclonal Antibody [clone: BID40]
Antibodies
Mouse monoclonal antibody raised against hepatitis B virus surface antigen (ad/ay).

Type: Primary
Antigen: Hepatitis B virus surface antigen (ad/ay)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: BID40
Epitope:
Host: IgG2b
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR