Anti-IMMT Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12412
10721-376EA 530.32 USD
10721-376
Anti-IMMT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant IMMT.

Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:2000

Type: Primary
Antigen: IMMT
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Cat, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR